missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Acetyl-4-phenylpiperidine hydrochloride, 99%, Thermo Scientific Chemicals
£145.00 - £503.00
Chemical Identifiers
CAS | 10315-03-4 |
---|---|
Molecular Formula | C13H17NO·ClH |
Molecular Weight (g/mol) | 239.74 |
MDL Number | MFCD00039037 |
InChI Key | JYDHZOIDIWUHDB-UHFFFAOYSA-N |
Synonym | 4-acetyl-4-phenylpiperidine hydrochloride, 1-4-phenylpiperidin-4-yl ethanone hydrochloride, 1-4-phenylpiperidin-4-yl ethan-1-one hydrochloride, 4-acetyl-4-phenylpiperidine, chloride, 1-4-phenylpiperidin-4-yl ethan-1-one hcl, acmc-1c3wz, 4-acetyl-4-phenylpiperidine hcl, 4-acetyl 4-phenyl piperidine hydrochloride, 4-acetyl-4-phenyl-piperidine hydrochloride |
PubChem CID | 2723767 |
IUPAC Name | 1-(4-phenylpiperidin-4-yl)ethanone;hydrochloride |
SMILES | CC(=O)C1(CCNCC1)C2=CC=CC=C2.Cl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10582531
|
Thermo Scientific Acros
134580050 |
5 g | Glass bottle |
£145.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10592721
|
Thermo Scientific Acros
134580250 |
25 g | Glass bottle |
£503.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10315-03-4 | |
239.74 | |
JYDHZOIDIWUHDB-UHFFFAOYSA-N | |
2723767 | |
CC(=O)C1(CCNCC1)C2=CC=CC=C2.Cl |
C13H17NO·ClH | |
MFCD00039037 | |
4-acetyl-4-phenylpiperidine hydrochloride, 1-4-phenylpiperidin-4-yl ethanone hydrochloride, 1-4-phenylpiperidin-4-yl ethan-1-one hydrochloride, 4-acetyl-4-phenylpiperidine, chloride, 1-4-phenylpiperidin-4-yl ethan-1-one hcl, acmc-1c3wz, 4-acetyl-4-phenylpiperidine hcl, 4-acetyl 4-phenyl piperidine hydrochloride, 4-acetyl-4-phenyl-piperidine hydrochloride | |
1-(4-phenylpiperidin-4-yl)ethanone;hydrochloride |
Specifications
10315-03-4 | |
100.0 | |
Yellow to Beige | |
99% | |
MFCD00039037 | |
JYDHZOIDIWUHDB-UHFFFAOYSA-N | |
1-(4-phenylpiperidin-4-yl)ethanone;hydrochloride | |
2723767 | |
99% | |
4-Acetyl-4-phenylpiperidine hydrochloride, 99% |
98.5 | |
232°C to 234°C | |
Authentic | |
C13H17NO·ClH | |
4-acetyl-4-phenylpiperidine hydrochloride, 1-4-phenylpiperidin-4-yl ethanone hydrochloride, 1-4-phenylpiperidin-4-yl ethan-1-one hydrochloride, 4-acetyl-4-phenylpiperidine, chloride, 1-4-phenylpiperidin-4-yl ethan-1-one hcl, acmc-1c3wz, 4-acetyl-4-phenylpiperidine hcl, 4-acetyl 4-phenyl piperidine hydrochloride, 4-acetyl-4-phenyl-piperidine hydrochloride | |
CC(=O)C1(CCNCC1)C2=CC=CC=C2.Cl | |
239.74 | |
239.74 | |
Powder |
Safety and Handling
missing translation for 'einecsNumber' : 233-694-0
RUO – Research Use Only