missing translation for 'onlineSavingsMsg'
Learn More

4-Acetylbenzenesulfonyl chloride, 97%, Thermo Scientific Chemicals

Product Code. 10064103
Click to view available options
Quantity:
1 g
5 g
50 g
Packaging:
Glass bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1788-10-9
Molecular Formula C8H7ClO3S
Molecular Weight (g/mol) 218.65
MDL Number MFCD00800269
InChI Key FXVDNCRTKXMSEZ-UHFFFAOYSA-N
Synonym 4-acetylbenzene-1-sulfonyl chloride, 4-acetyl-benzenesulfonyl chloride, 4-acetylphenylsulfonyl chloride, benzenesulfonylchloride, 4-acetyl, benzenesulfonyl chloride, 4-acetyl, 1-acetyl-4-chlorosulfonyl benzene, 4-acetylbenzenesulfonyl chloride, acmc-1bs75, ksc183k3b, 4-acetylbenzenesulfonylchloride
PubChem CID 266418
IUPAC Name 4-acetylbenzenesulfonyl chloride
SMILES CC(=O)C1=CC=C(C=C1)S(=O)(=O)Cl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 83°C to 87°C
Color Brown to White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula H3CCO(C6H4)SO2Cl
Quantity 50 g
Formula Weight 218.65
Percent Purity 97%
Physical Form Chunks and/or Powder
Chemical Name or Material 4-Acetylbenzenesulfonyl chloride, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Acetylbenzenesulfonyl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 C
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'