missing translation for 'onlineSavingsMsg'
Learn More

4-Acetylphenoxyacetic acid, 98+%, Thermo Scientific™

£31.20 - £202.00

Chemical Identifiers

CAS 1878-81-5
Molecular Formula C10H10O4
Molecular Weight (g/mol) 194.19
MDL Number MFCD00014363
InChI Key KMXZEXUYXUMHEQ-UHFFFAOYSA-N
Synonym 4-acetylphenoxyacetic acid, 2-4-acetylphenoxy acetic acid, 4-acetylphenoxy acetic acid, p-acetylphenoxy acetic acid, p-acetylphenoxyacetic acid, chembl84623, 4-acetyl-phenoxy-acetic acid, acetic acid, 4-acetylphenoxy, acetic acid,2-4-acetylphenoxy
PubChem CID 74655
IUPAC Name 2-(4-acetylphenoxy)acetic acid
SMILES CC(=O)C1=CC=C(OCC(O)=O)C=C1
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11350034
View Documents
Thermo Scientific Alfa Aesar
L00426.06
5 g
£31.20
5g
Please to purchase this item. Need a web account? Register with us today!
11360034
View Documents
Thermo Scientific Alfa Aesar
L00426.14
25 g
£99.10
25g
Please to purchase this item. Need a web account? Register with us today!
11370034
View Documents
Thermo Scientific Alfa Aesar
L00426.22
100 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

1878-81-5
194.19
KMXZEXUYXUMHEQ-UHFFFAOYSA-N
74655
CC(=O)C1=CC=C(OCC(O)=O)C=C1
C10H10O4
MFCD00014363
4-acetylphenoxyacetic acid, 2-4-acetylphenoxy acetic acid, 4-acetylphenoxy acetic acid, p-acetylphenoxy acetic acid, p-acetylphenoxyacetic acid, chembl84623, 4-acetyl-phenoxy-acetic acid, acetic acid, 4-acetylphenoxy, acetic acid,2-4-acetylphenoxy
2-(4-acetylphenoxy)acetic acid

Specifications

1878-81-5
C10H10O4
1956136
KMXZEXUYXUMHEQ-UHFFFAOYSA-N
2-(4-acetylphenoxy)acetic acid
74655
≥98%
172°C to 174°C
MFCD00014363
4-acetylphenoxyacetic acid, 2-4-acetylphenoxy acetic acid, 4-acetylphenoxy acetic acid, p-acetylphenoxy acetic acid, p-acetylphenoxyacetic acid, chembl84623, 4-acetyl-phenoxy-acetic acid, acetic acid, 4-acetylphenoxy, acetic acid,2-4-acetylphenoxy
CC(=O)C1=CC=C(OCC(O)=O)C=C1
194.19
194.19
4-Acetylphenoxyacetic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'einecsNumber' : 217-524-2

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications