missing translation for 'onlineSavingsMsg'
Learn More

4-Allyloxybenzoyl chloride, 98%, Thermo Scientific™

Product Code. 10197153
1 g, Glass bottle
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

CAS 36844-51-6
CAS Min % 97.5
CAS Max % 100.0
Density 1.1550g/mL
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (Argentometry)
Packaging Glass bottle
Molecular Formula C10H9ClO2
Linear Formula CH2CHCH2OC6H4COCl
MDL Number MFCD09744024
Quantity 1 g
Specific Gravity 1.155
Synonym 4-allyloxybenzoyl chloride, benzoyl chloride, 4-2-propen-1-yloxy, 4-prop-2-en-1-yloxy benzoyl chloride, p-allyloxybenzoyl chloride, 4-allyloxy benzoyl chloride, 4-2-propenyloxy benzoyl chloride, 4-allyloxy benzoic acid chloride, 4-prop-2-en-1-oxy benzoyl chloride, benzoyl chloride,4-2-propen-1-yloxy
InChI Key WYMVIVUKHYHNBN-UHFFFAOYSA-N
SMILES C=CCOC1=CC=C(C=C1)C(=O)Cl
IUPAC Name 4-prop-2-enoxybenzoyl chloride
Molecular Weight (g/mol) 196.63
PubChem CID 11790137
Formula Weight 196.63
Percent Purity 98%
Physical Form Low Melting Solid
Chemical Name or Material 4-Allyloxybenzoyl chloride
Show More Show Less
compliance-icons
Product Identifier
  • 4-Allyloxybenzoyl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Allyloxybenzoyl chloride, 98%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.