missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-2-chlorobenzoic acid, 98%, Thermo Scientific Chemicals

Product Code. 11309439
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2457-76-3
Molecular Formula C7H6ClNO2
Molecular Weight (g/mol) 171.58
MDL Number MFCD00007772
InChI Key MBDUKNCPOPMRJQ-UHFFFAOYSA-N
Synonym 2-chloro-4-aminobenzoic acid, benzoic acid, 4-amino-2-chloro, o-chloro-p-aminobenzoic acid, 2-chloro-p-aminobenzoic acid, usaf nb-1, 4-amino-2-chloro-benzoic acid, 4-amino-2-chlorobenzoicacid, unii-y3s6924ia6, wln: zr cg dvq, pubchem3109
PubChem CID 17154
ChEBI CHEBI:59472
IUPAC Name 4-amino-2-chlorobenzoic acid
SMILES C1=CC(=C(C=C1N)Cl)C(=O)O

Specifications

Melting Point ∼220°C (decomposition)
Quantity 100 g
Beilstein 2803668
Formula Weight 171.58
Percent Purity 98%
Chemical Name or Material 4-Amino-2-chlorobenzoic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'