missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-2-chlorobenzoic acid, 98%, Thermo Scientific Chemicals

£41.66 - £507.53

Chemical Identifiers

CAS 2457-76-3
Molecular Formula C7H6ClNO2
Molecular Weight (g/mol) 171.58
MDL Number MFCD00007772
InChI Key MBDUKNCPOPMRJQ-UHFFFAOYSA-N
Synonym 2-chloro-4-aminobenzoic acid, benzoic acid, 4-amino-2-chloro, o-chloro-p-aminobenzoic acid, 2-chloro-p-aminobenzoic acid, usaf nb-1, 4-amino-2-chloro-benzoic acid, 4-amino-2-chlorobenzoicacid, unii-y3s6924ia6, wln: zr cg dvq, pubchem3109
PubChem CID 17154
ChEBI CHEBI:59472
IUPAC Name 4-amino-2-chlorobenzoic acid
SMILES C1=CC(=C(C=C1N)Cl)C(=O)O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11459362
View Documents
Thermo Scientific Alfa Aesar
A10116.06
5 g
£41.66
5g
Please to purchase this item. Need a web account? Register with us today!
11469362
View Documents
Thermo Scientific Alfa Aesar
25 g
£133.67
25g
Please to purchase this item. Need a web account? Register with us today!
11309439
View Documents
Thermo Scientific Alfa Aesar
100 g
£507.53
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

2457-76-3
171.58
MBDUKNCPOPMRJQ-UHFFFAOYSA-N
17154
4-amino-2-chlorobenzoic acid
C7H6ClNO2
MFCD00007772
2-chloro-4-aminobenzoic acid, benzoic acid, 4-amino-2-chloro, o-chloro-p-aminobenzoic acid, 2-chloro-p-aminobenzoic acid, usaf nb-1, 4-amino-2-chloro-benzoic acid, 4-amino-2-chlorobenzoicacid, unii-y3s6924ia6, wln: zr cg dvq, pubchem3109
CHEBI:59472
C1=CC(=C(C=C1N)Cl)C(=O)O

Specifications

2457-76-3
C7H6ClNO2
2803668
MBDUKNCPOPMRJQ-UHFFFAOYSA-N
4-amino-2-chlorobenzoic acid
17154
171.58
4-Amino-2-chlorobenzoic acid
∼220°C (decomposition)
MFCD00007772
2-chloro-4-aminobenzoic acid, benzoic acid, 4-amino-2-chloro, o-chloro-p-aminobenzoic acid, 2-chloro-p-aminobenzoic acid, usaf nb-1, 4-amino-2-chloro-benzoic acid, 4-amino-2-chlorobenzoicacid, unii-y3s6924ia6, wln: zr cg dvq, pubchem3109
C1=CC(=C(C=C1N)Cl)C(=O)O
171.58
CHEBI:59472
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'einecsNumber' : 219-540-5

missing translation for 'rtecsNumber' : DG1575000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only