missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-2-nitrophenol, 99%

Product Code. 10408900 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Unit Size:
5g
This item is not returnable. View return policy

Product Code. 10408900

Brand: Thermo Scientific Acros 164230050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 119-34-6 | C6H6N2O3 | 154.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 119-34-6
CAS Min % 98.5
CAS Max % 100.0
Melting Point 124.0°C to 128.0°C
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Molecular Formula C6H6N2O3
Linear Formula H2NC6H3(NO2)OH
MDL Number MFCD00007876
Quantity 5 g
Beilstein 13, 520
Synonym phenol, 4-amino-2-nitro, fourrine 57, 4-hydroxy-3-nitroaniline, 2-nitro-4-aminophenol, fourrine brown pr, oxidation base 25, fourrine brown propyl, o-nitro-p-aminophenol, p-amino-o-nitrophenol, c.i. oxidation base 25
Solubility Information Solubility in water: .. Other solubilities: soluble in diethylether, ethanol and aceton
InChI Key WHODQVWERNSQEO-UHFFFAOYSA-N
SMILES C1=CC(=C(C=C1N)[N+](=O)[O-])O
IUPAC Name 4-amino-2-nitrophenol
Molecular Weight (g/mol) 154.12
PubChem CID 3417419
ChEBI CHEBI:82385
Formula Weight 154.12
Percent Purity 99%
Chemical Name or Material 4-Amino-2-nitrophenol, 99%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 4-Amino-2-nitrophenol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Germ cell mutagenicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
  • H341-Suspected of causing genetic defects.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P362-Take off contaminated clothing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.