missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-3-chlorophenol hydrochloride, 98%, Thermo Scientific™

£31.21 - £97.81

Chemical Identifiers

CAS 52671-64-4
Molecular Formula C6H6ClNO·ClH
Molecular Weight (g/mol) 180.03
MDL Number MFCD00143110
InChI Key RFJVQGMBFQGZPV-UHFFFAOYSA-N
Synonym 4-amino-3-chlorophenol hydrochloride, 4-amino-3-chlorophenol hcl, 2-chloro-4-hydroxyaniline hydrochloride, phenol, 4-amino-3-chloro-, hydrochloride, 4-azanyl-3-chloranyl-phenol hydrochloride, c6h6clno.hcl, acmc-209l0t, ksc495i8f, 4-amino-3-chlorophenol, chloride, 3-chloro-4-aminophenol hydrochloride
PubChem CID 12598190
IUPAC Name 4-amino-3-chlorophenol;hydrochloride
SMILES C1=CC(=C(C=C1O)Cl)N.Cl
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10274584
SDS View Documents
Thermo Scientific Acros
399310050
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10234634
SDS View Documents
Thermo Scientific Acros
399310250
25 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Specifications
Show More
Videos
Safety and Handling
Show More
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Amino-3-chlorophenol hydrochloride, 98%, Thermo Scientific™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.