missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-3-fluorobenzoic acid, 98%, Thermo Scientific Chemicals

Product Code. 15413377
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 455-87-8
Molecular Formula C7H6FNO2
Molecular Weight (g/mol) 155.128
MDL Number MFCD01660374
InChI Key JSKXHTHMCCDEGD-UHFFFAOYSA-N
Synonym 3-fluoro-4-aminobenzoic acid, benzoic acid, 4-amino-3-fluoro, 4-amino-3-fluorobenzenecarboxylic acid, 4-amino-3-fluoro-benzoic acid, 4-amino-3-fluorobenzoicacid, pubchem3524, wln: zr bf dvq, acmc-209k3i, 3-14-00-01153 beilstein handbook reference, buttpark 44\09-60
PubChem CID 9971
IUPAC Name 4-amino-3-fluorobenzoic acid
SMILES C1=CC(=C(C=C1C(=O)O)F)N

Specifications

Melting Point 214°C to 216°C
Quantity 250 mg
Formula Weight 155.13
Percent Purity 98%
Chemical Name or Material 4-Amino-3-fluorobenzoic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'