Learn More
4-Amino-3-methylphenol, 98%, Thermo Scientific Chemicals
£34.07 - £93.06
Chemical Identifiers
CAS | 2835-99-6 |
---|---|
Molecular Formula | C7H9NO |
Molecular Weight (g/mol) | 123.155 |
MDL Number | MFCD00007871 |
InChI Key | QGNGOGOOPUYKMC-UHFFFAOYSA-N |
Synonym | 4-amino-m-cresol, 4-hydroxy-2-methylaniline, 3-methyl-4-aminophenol, p-amino-m-cresol, phenol, 4-amino-3-methyl, p-hydroxy-o-toluidine, 2-methyl-4-hydroxyaniline, 2-amino-5-hydroxytoluene, m-cresol, 4-amino, 4-amino-3-cresol |
PubChem CID | 17819 |
ChEBI | CHEBI:55546 |
IUPAC Name | 4-amino-3-methylphenol |
SMILES | CC1=C(C=CC(=C1)O)N |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11416386
|
Thermo Scientific Alfa Aesar
B23152.14 |
25 g |
£34.07
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11426386
|
Thermo Scientific Alfa Aesar
B23152.22 |
100 g |
£93.06
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
2835-99-6 | |
123.155 | |
QGNGOGOOPUYKMC-UHFFFAOYSA-N | |
17819 | |
4-amino-3-methylphenol |
C7H9NO | |
MFCD00007871 | |
4-amino-m-cresol, 4-hydroxy-2-methylaniline, 3-methyl-4-aminophenol, p-amino-m-cresol, phenol, 4-amino-3-methyl, p-hydroxy-o-toluidine, 2-methyl-4-hydroxyaniline, 2-amino-5-hydroxytoluene, m-cresol, 4-amino, 4-amino-3-cresol | |
CHEBI:55546 | |
CC1=C(C=CC(=C1)O)N |
Specifications
2835-99-6 | |
C7H9NO | |
UN3077 | |
4-amino-m-cresol, 4-hydroxy-2-methylaniline, 3-methyl-4-aminophenol, p-amino-m-cresol, phenol, 4-amino-3-methyl, p-hydroxy-o-toluidine, 2-methyl-4-hydroxyaniline, 2-amino-5-hydroxytoluene, m-cresol, 4-amino, 4-amino-3-cresol | |
CC1=C(C=CC(=C1)O)N | |
123.155 | |
CHEBI:55546 | |
98% |
176°C to 179°C | |
MFCD00007871 | |
2078803 | |
QGNGOGOOPUYKMC-UHFFFAOYSA-N | |
4-amino-3-methylphenol | |
17819 | |
123.16 | |
4-Amino-3-methylphenol |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
missing translation for 'dotInformation' : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
missing translation for 'rtecsNumber' : GO6826000
missing translation for 'tsca' : TSCA
RUO – Research Use Only