missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-5-imidazolecarboxamide hydrochloride, 98%, Thermo Scientific Chemicals

£16.16 - £195.00

Chemical Identifiers

CAS 72-40-2, 360-97-4
Molecular Formula C4H7ClN4O
Molecular Weight (g/mol) 162.58
MDL Number MFCD00012704
InChI Key MXCUYSMIELHIQL-UHFFFAOYSA-N
Synonym 4-amino-5-imidazolecarboxamide hydrochloride, 5-amino-4-imidazolecarboxamide hydrochloride, 5-amino-1h-imidazole-4-carboxamide hydrochloride, 5-aminoimidazole-4-carboxamide hydrochloride, aic .cntdot. hcl, 4-amino-1h-imidazole-5-carboxamide hydrochloride, aminoimidazolecarboxamide hydrochloride, 4-aminoimidazole-5-carboxamide hydrochloride, 1h-imidazole-4-carboxamide, 5-amino-, monohydrochloride, 5-aminoimidazol-4-carboxamide, hydrochloride
PubChem CID 66146
IUPAC Name 4-amino-1H-imidazole-5-carboxamide;hydrochloride
SMILES Cl.NC(=O)C1=C(N)N=CN1
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10405295
View Documents
Thermo Scientific Acros
173090010
1 g Glass bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10275940
View Documents
Thermo Scientific Acros
173090050
5 g Glass bottle
£57.60
5g
Please to purchase this item. Need a web account? Register with us today!
10684852
View Documents
Thermo Scientific Acros
173090250
25 g Glass bottle
£195.00
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

72-40-2, 360-97-4
162.58
MXCUYSMIELHIQL-UHFFFAOYSA-N
66146
Cl.NC(=O)C1=C(N)N=CN1
C4H7ClN4O
MFCD00012704
4-amino-5-imidazolecarboxamide hydrochloride, 5-amino-4-imidazolecarboxamide hydrochloride, 5-amino-1h-imidazole-4-carboxamide hydrochloride, 5-aminoimidazole-4-carboxamide hydrochloride, aic .cntdot. hcl, 4-amino-1h-imidazole-5-carboxamide hydrochloride, aminoimidazolecarboxamide hydrochloride, 4-aminoimidazole-5-carboxamide hydrochloride, 1h-imidazole-4-carboxamide, 5-amino-, monohydrochloride, 5-aminoimidazol-4-carboxamide, hydrochloride
4-amino-1H-imidazole-5-carboxamide;hydrochloride

Specifications

72-40-2, 360-97-4
100.0
White
0.5% max. (105°C)
97.5% min. (Argentometry)
MFCD00012704
4-amino-5-imidazolecarboxamide hydrochloride, 5-amino-4-imidazolecarboxamide hydrochloride, 5-amino-1h-imidazole-4-carboxamide hydrochloride, 5-aminoimidazole-4-carboxamide hydrochloride, aic .cntdot. hcl, 4-amino-1h-imidazole-5-carboxamide hydrochloride, aminoimidazolecarboxamide hydrochloride, 4-aminoimidazole-5-carboxamide hydrochloride, 1h-imidazole-4-carboxamide, 5-amino-, monohydrochloride, 5-aminoimidazol-4-carboxamide, hydrochloride
MXCUYSMIELHIQL-UHFFFAOYSA-N
4-amino-1H-imidazole-5-carboxamide;hydrochloride
66146
98%
4-Amino-5-imidazolecarboxamide hydrochloride, 98%
97.5
250°C
0.2% max.
Authentic
C4H7ClN4O
25, II, 221
(5% in water) Clear
Cl.NC(=O)C1=C(N)N=CN1
162.58
162.58
Crystalline Powder or Crystals
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 200-778-3

missing translation for 'rtecsNumber' : NI3911000

missing translation for 'tsca' : TSCA

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.