missing translation for 'onlineSavingsMsg'
Learn More

4-Aminoantipyrine, 97%, Thermo Scientific Chemicals

Product Code. 11401508
Click to view available options
Quantity:
100 g
500 g
2500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

4-Aminoantipyrine shows both analgesic and anti-inflammatory properties. It is used as a reagent for glucose determination in the presence of peroxidase and phenol. It is used as a drug dipyrone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Aminoantipyrine shows both analgesic and anti-inflammatory properties. It is used as a reagent for glucose determination in the presence of peroxidase and phenol. It is used as a drug dipyrone.

Solubility
Soluble in water. 500 g/L (20°C), methanol. Partially soluble in diethyl ether.

Notes
Incompatible with Strong oxidizing agents, strong acids, acid chlorides, acid anhydrides.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 83-07-8
Molecular Formula C11H13N3O
Molecular Weight (g/mol) 203.245
MDL Number MFCD00003145
InChI Key RLFWWDJHLFCNIJ-UHFFFAOYSA-N
Synonym 4-aminoantipyrine, ampyrone, metapirazone, aminoantipyrine, solvapyrin-a, aminoazophene, 4-aminophenazone, 4-aminoantipyrene, aminoantipyrin, solnapyrin-a
PubChem CID 2151
ChEBI CHEBI:59026
IUPAC Name 4-amino-1,5-dimethyl-2-phenylpyrazol-3-one
SMILES CC1=C(C(=O)N(N1C)C2=CC=CC=C2)N

Specifications

Melting Point 106°C to 110°C
Assay Percent Range 97%
Quantity 500 g
Beilstein 181635
Merck Index 14,591
Solubility Information Soluble in water. 500g/L (20°C),methanol. Partially soluble in diethyl ether.
Formula Weight 203.25
Percent Purity 97%
Chemical Name or Material 4-Aminoantipyrine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'