Learn More
4-Aminoantipyrine, 98%, Thermo Scientific Chemicals
4-Aminoantipyrine, 98%, C11H13N3O, CAS Number-83-07-8, 4-aminoantipyrene, aminoantipyrin, ampyrone, 4-aminoantipyrine, 4-aminophenazone, metapirazone, solnapyrin-a, aminoazophene, solvapyrin-a, aminoantipyrine, 25g, (5% aq. soln.) clear, 24,273, 100.0, 97.5, CHEBI:59026, Brown-Yellow to Yellow
£30.00 - £1008.00
Chemical Identifiers
CAS | 83-07-8 |
---|---|
Molecular Formula | C11H13N3O |
Molecular Weight (g/mol) | 203.25 |
MDL Number | MFCD00003145 |
InChI Key | RLFWWDJHLFCNIJ-UHFFFAOYSA-N |
Synonym | 4-aminoantipyrine, ampyrone, metapirazone, aminoantipyrine, solvapyrin-a, aminoazophene, 4-aminophenazone, 4-aminoantipyrene, aminoantipyrin, solnapyrin-a |
PubChem CID | 2151 |
ChEBI | CHEBI:59026 |
IUPAC Name | 4-amino-1,5-dimethyl-2-phenylpyrazol-3-one |
SMILES | CC1=C(C(=O)N(N1C)C2=CC=CC=C2)N |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10795161
|
Thermo Scientific Acros
103150250 |
25 g | Glass bottle |
£30.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10497660
|
Thermo Scientific Acros
103151000 |
100 g | Glass bottle |
£79.40
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10705171
|
Thermo Scientific Acros
103155000 |
500 g | Glass bottle |
£261.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11946863
|
Thermo Scientific Acros
103150025 |
2.5 kg | Plastic Bottle |
£1008.00
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
83-07-8 | |
203.25 | |
RLFWWDJHLFCNIJ-UHFFFAOYSA-N | |
2151 | |
4-amino-1,5-dimethyl-2-phenylpyrazol-3-one |
C11H13N3O | |
MFCD00003145 | |
4-aminoantipyrine, ampyrone, metapirazone, aminoantipyrine, solvapyrin-a, aminoazophene, 4-aminophenazone, 4-aminoantipyrene, aminoantipyrin, solnapyrin-a | |
CHEBI:59026 | |
CC1=C(C(=O)N(N1C)C2=CC=CC=C2)N |
Specifications
83-07-8 | |
100.0 | |
Brown-Yellow to Yellow | |
1.5% max. | |
98% | |
MFCD00003145 | |
15,413 | |
4-aminoantipyrine, ampyrone, metapirazone, aminoantipyrine, solvapyrin-a, aminoazophene, 4-aminophenazone, 4-aminoantipyrene, aminoantipyrin, solnapyrin-a | |
RLFWWDJHLFCNIJ-UHFFFAOYSA-N | |
4-amino-1,5-dimethyl-2-phenylpyrazol-3-one | |
2151 | |
203.25 | |
Crystals or Powder |
97.5 | |
105.5°C to 110.0°C | |
0.8000g/mL | |
Authentic | |
C11H13N3O | |
24,273 | |
0.8 | |
(5% in water) Clear | |
CC1=C(C(=O)N(N1C)C2=CC=CC=C2)N | |
203.25 | |
CHEBI:59026 | |
98% | |
4-Aminoantipyrine, 98% |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attent
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 201-452-3
missing translation for 'rtecsNumber' : CD2480000
missing translation for 'tsca' : TSCA
- 4-Aminoantipyrine
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only