missing translation for 'onlineSavingsMsg'
Learn More

4-Aminobenzo-2,1,3-thiadiazole, 98%

Product Code. 11302207
1 g, 1g
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11302207

Brand: Thermo Scientific Alfa Aesar L09678.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 767-64-6 | C6H5N3S | 151.187 g/mol

4-Aminobenzo-2,1,3-thiadiazole is used in a study to synthesize and evaluate the inhibitory activity of a series of substituted benzimidazoles and small benzothiadiazoles on rat liver methionine synthase. It was used as potential levelers for Cu plating in submicrometer trenches.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Aminobenzo-2,1,3-thiadiazole is used in a study to synthesize and evaluate the inhibitory activity of a series of substituted benzimidazoles and small benzothiadiazoles on rat liver methionine synthase. It was used as potential levelers for Cu plating in submicrometer trenches.

Solubility
Insoluble in water.

Notes
Incompatible with oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 767-64-6
Molecular Formula C6H5N3S
Molecular Weight (g/mol) 151.187
MDL Number MFCD00005810
InChI Key DRLGIZIAMHIQHL-UHFFFAOYSA-N
Synonym 4-aminobenzo-2,1,3-thiadiazole, 4-amino-2,1,3-benzothiadiazole, 4-aminopiazthiole, benzo c 1,2,5 thiadiazol-4-amine, 2,1,3-benzothiadiazole, 4-amino, 7-amino-2,1,3-benzothiadiazole, benzo 1,2,5 thiadiazol-4-ylamine, 2,1,3-benzothiadiazole, 7-amino, 2,1,3-benzothiadiazol-4-ylamine, 2,1,3-benzothiadiazol-4 1h-imine
PubChem CID 69845
IUPAC Name 2,1,3-benzothiadiazol-4-amine
SMILES C1=CC2=NSN=C2C(=C1)N

Specifications

Melting Point 67°C to 69°C
Quantity 1 g
Beilstein 3551
Solubility Information Insoluble in water.
Formula Weight 151.19
Percent Purity 98%
Chemical Name or Material 4-Aminobenzo-2,1,3-thiadiazole

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Product Code
Lot Number10245572 Certificate TypeCertificate of Analysis Date01/02/2023 Product CodeL09678.06, L09678.03
Lot Number0.101104 Certificate TypeCertificate of Analysis Date08/12/2021 Product CodeL09678.06, L09678.03
Lot Number10148551 Certificate TypeCertificate of Analysis Date08/12/2021 Product CodeL09678.06, L09678.03
Lot Number10196736 Certificate TypeCertificate of Analysis Date07/12/2021 Product CodeL09678.06, L09678.03
Lot Number10111426 Certificate TypeCertificate of Analysis Date07/12/2021 Product CodeL09678.06, L09678.03

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Aminobenzo-2,1,3-thiadiazole, 98% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.