missing translation for 'onlineSavingsMsg'
Learn More

4-Aminobutyric acid, 99+%, Thermo Scientific Chemicals

Product Code. 15129153
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Plastic Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

4-Aminobutyric acid, 99+%, CAS # 56-12-2, also known as γ-Aminobutyric acid, is an amino acid that shows inhibitory activities against neurotransmitters, including GABA.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• 4-Aminobutyric acid is a γ-amino acid and monocarboxylic acid derived from butyric acid

• It is an amino acid that shows inhibitory activities against neurotransmitters, including GABA, and acts by binding GABA-A receptor and inhibiting the activity of signal-receiving neurons

• This compound, converted from neurotransmitter glutamate, is essential to regulate neuronal excitability

• It binds to GABA-A and GABA-B receptors, causing ion channel opening, hyperpolarization, and eventually inhibition of neurotransmission

Applications

• 4-Aminobutyric acid has been used in the preparation of GABA–BSA (bovine serum albumin)–GA (glutaraldehyde) conjugate, which is used as a control for immunohistochemical experiments

• It is used, in vivo, to detect GABA neurotransmitter by magnetic resonance spectroscopy

• This compound increases the production of short-chain fatty acids and decreases pH values in mouse colon

• Some in vivo experiments, using rats, mice, and pigs, demonstrated that 4-Aminobutyric acid can modulate intestinal fluid secretion, blood flow and ulceration

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-12-2
Molecular Formula C4H9NO2
Molecular Weight (g/mol) 103.12
MDL Number MFCD00008226
InChI Key BTCSSZJGUNDROE-UHFFFAOYSA-N
Synonym 4-aminobutyric acid, gaba, gamma-aminobutyric acid, piperidic acid, piperidinic acid, aminalon, gaballon, gammalon, mielogen, butanoic acid, 4-amino
PubChem CID 119
ChEBI CHEBI:16865
IUPAC Name 4-aminobutanoic acid
SMILES C(CC(=O)O)CN

Specifications

Melting Point 197°C to 204°C
CAS Min % 99.0
CAS Max % 100.0
Color White
Infrared Spectrum Authentic
Assay Percent Range 99+%
Loss on Drying 0.5% max.
Packaging Plastic Bottle
Linear Formula H2N(CH2)3CO2H
Beilstein 04,413
Merck Index 15,425
Transmittance 95% min. (c=10, 2N HCl, 430nm) 1cm cell
Solubility Information Solubility in water: very soluble. Other solubilities: slightly soluble in common solvents
Quantity 2.5 kg
Formula Weight 103.12
Percent Purity ≥99%
Physical Form Crystalline Powder
Chemical Name or Material 4-Aminobutyric acid, 99+%
Show More Show Less
Product Identifier
  • 4-Aminobutyric acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.