missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Aminohippuric Acid EU Pharmacopoeia, Fisher Chemical™
Click to view available options
Quantity:
100 mL
Packaging:
Bottle
Specifications
CAS | 66003-55-2 |
Packaging | Bottle |
Quantity | 100 mL |
Type | Mass Spectrometry Standard |
Grade | Eur Ph Ref.: 1003701 |
Solvent or Matrix | Methanol |
Chemical Name or Material | Aminohippuric acid reagent. |
Safety and Handling
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS02_Extremely_Flammable.png)
Product Identifier
- Aminohippuric acid reagent. European Pharmacopoeia Ref: 1003701. (RP 009)
- Danger
- Flammable liquid Category 2
- H225-Highly flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P233-Keep container tightly closed.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P370+P378-In case of fire: Use to extinguish.
- P403+P235-Store in a well-ventilated place. Keep cool.
- P501-Dispose of contents/container to .
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'