Learn More
4-Aminophenyl ether, 98%, Thermo Scientific Chemicals
£22.10 - £53.60
Chemical Identifiers
CAS | 101-80-4 |
---|---|
Molecular Formula | C12H12N2O |
Molecular Weight (g/mol) | 200.24 |
MDL Number | MFCD00007863 |
InChI Key | HLBLWEWZXPIGSM-UHFFFAOYSA-N |
Synonym | 4,4'-oxydianiline, 4,4'-diaminodiphenyl ether, 4,4-oxydianiline, 4-aminophenyl ether, 4,4'-oxybisbenzenamine, oxydianiline, 4-4-aminophenoxy aniline, diaminodiphenyl ether, benzenamine, 4,4'-oxybis, p,p'-oxydianiline |
PubChem CID | 7579 |
ChEBI | CHEBI:34384 |
IUPAC Name | 4-(4-aminophenoxy)aniline |
SMILES | C1=CC(=CC=C1N)OC2=CC=C(C=C2)N |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10459200
|
Thermo Scientific Acros
104330050 |
5 g | Glass bottle | ||||||
10611752
|
Thermo Scientific Acros
104330250 |
25 g | Glass bottle |
£25.80
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10434915
|
Thermo Scientific Acros
104331000 |
100 g | Glass bottle |
£53.60
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
101-80-4 | |
200.24 | |
HLBLWEWZXPIGSM-UHFFFAOYSA-N | |
7579 | |
4-(4-aminophenoxy)aniline |
C12H12N2O | |
MFCD00007863 | |
4,4'-oxydianiline, 4,4'-diaminodiphenyl ether, 4,4-oxydianiline, 4-aminophenyl ether, 4,4'-oxybisbenzenamine, oxydianiline, 4-4-aminophenoxy aniline, diaminodiphenyl ether, benzenamine, 4,4'-oxybis, p,p'-oxydianiline | |
CHEBI:34384 | |
C1=CC(=CC=C1N)OC2=CC=C(C=C2)N |
Specifications
101-80-4 | |
100.0 | |
190°C (0.1 mmHg) | |
98% | |
(H2NC6H4)2O | |
13, 441 | |
Solubility in water: negligible | |
C1=CC(=CC=C1N)OC2=CC=C(C=C2)N | |
200.24 | |
CHEBI:34384 | |
98% |
97.5 | |
189°C to 193°C | |
218°C | |
C12H12N2O | |
MFCD00007863 | |
4,4'-oxydianiline, 4,4'-diaminodiphenyl ether, 4,4-oxydianiline, 4-aminophenyl ether, 4,4'-oxybisbenzenamine, oxydianiline, 4-4-aminophenoxy aniline, diaminodiphenyl ether, benzenamine, 4,4'-oxybis, p,p'-oxydianiline | |
HLBLWEWZXPIGSM-UHFFFAOYSA-N | |
4-(4-aminophenoxy)aniline | |
7579 | |
200.24 | |
4-Aminophenyl ether |
Safety and Handling
GHS H Statement:
May cause cancer.
May cause genetic defects.
Suspected of damaging fertility.
Toxic if inhaled.
Toxic in contact with skin.
Toxic if swallowed.
Toxic to aquatic life with long lasting effects.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF exposed or concerned: Get medical advice/attention.
Avoid release to the environment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 202-977-
- 4-Aminophenyl ether
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 2
- Carcinogenicity Category 1B
- Germ cell mutagenicity Category 1B
- Reproductive toxicity Category 2
Hazard Statement
- H340-May cause genetic defects.
- H350-May cause cancer.
- H361f-Suspected of damaging fertility.
- H411-Toxic to aquatic life with long lasting effects.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H301-Toxic if swallowed.
Precautionary Statement
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
RUO – Research Use Only