missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4'-Aminopropiophenone, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | 135.0°C to 140.0°C |
Assay Percent Range | 98% |
Packaging | Glass bottle |
Linear Formula | NH2C6H4C(O)CH2CH3 |
Quantity | 10 g |
Merck Index | 15,466 |
Solubility Information | Solubility in water: soluble. Other solubilities: slightly soluble in ethanol, insoluble in diethyl ether |
Formula Weight | 149.19 |
Percent Purity | 98% |
Chemical Name or Material | 4'-Aminopropiophenone |
Sicurezza e movimentazione
Identificativo prodotto
- 4'-Aminopropiophenone
- Danger
- Acute toxicity Category 3
- H301-Toxic if swallowed.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'