Learn More
4-Aminothiophenol, 97%, Thermo Scientific Chemicals
£27.60 - £308.00
Chemical Identifiers
CAS | 1193-02-8 |
---|---|
Molecular Formula | C6H6NS |
Molecular Weight (g/mol) | 124.18 |
MDL Number | MFCD00007880 |
InChI Key | WCDSVWRUXWCYFN-UHFFFAOYSA-M |
Synonym | 4-aminothiophenol, 4-mercaptoaniline, p-aminothiophenol, p-aminobenzenethiol, 4-amino thiophenol, benzenethiol, 4-amino, p-mercaptoaniline, benzenethiol, p-amino, p-aminophenylmercaptan, 4-aminophenyl mercaptan |
PubChem CID | 14510 |
IUPAC Name | 4-aminobenzenethiol |
SMILES | NC1=CC=C([S-])C=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11408026
|
Thermo Scientific Alfa Aesar
A14082.06 |
5 g |
£27.60
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11418026
|
Thermo Scientific Alfa Aesar
A14082.14 |
25 g |
£94.10
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11428026
|
Thermo Scientific Alfa Aesar
A14082.22 |
100 g |
£308.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
4-Aminothiophenol is used in the synthesis of pectin-4-aminothiophenol conjugates and it forms self-assembled monolayers (SAMs) in the development of DNA. It is used in the detection of thrombin by protein recognition using surface-enhanced Raman spectroscopy (SERS), which finds applications in silver and gold nano particles. It acts as a linker to thiol (SH) functionalizes multiwalled carbon nanotubes (MWCNTs).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Aminothiophenol is used in the synthesis of pectin-4-aminothiophenol conjugates and it forms self-assembled monolayers (SAMs) in the development of DNA. It is used in the detection of thrombin by protein recognition using surface-enhanced Raman spectroscopy (SERS), which finds applications in silver and gold nano particles. It acts as a linker to thiol (SH) functionalizes multiwalled carbon nanotubes (MWCNTs).
Solubility
Immiscible with water.
Notes
Air sensitive. Store in a cool place. Incompatible with acids, acid chlorides, acid anhydrides, chloroformates and strong oxidizing agents.
Chemical Identifiers
1193-02-8 | |
124.18 | |
WCDSVWRUXWCYFN-UHFFFAOYSA-M | |
14510 | |
NC1=CC=C([S-])C=C1 |
C6H6NS | |
MFCD00007880 | |
4-aminothiophenol, 4-mercaptoaniline, p-aminothiophenol, p-aminobenzenethiol, 4-amino thiophenol, benzenethiol, 4-amino, p-mercaptoaniline, benzenethiol, p-amino, p-aminophenylmercaptan, 4-aminophenyl mercaptan | |
4-aminobenzenethiol |
Specifications
1193-02-8 | |
156°C to 157°C | |
97% | |
MFCD00007880 | |
906904 | |
4-aminothiophenol, 4-mercaptoaniline, p-aminothiophenol, p-aminobenzenethiol, 4-amino thiophenol, benzenethiol, 4-amino, p-mercaptoaniline, benzenethiol, p-amino, p-aminophenylmercaptan, 4-aminophenyl mercaptan | |
WCDSVWRUXWCYFN-UHFFFAOYSA-M | |
4-aminobenzenethiol | |
14510 | |
97% | |
4-Aminothiophenol |
37°C to 42°C | |
>110°C (230°F) | |
C6H6NS | |
UN3263 | |
Air Sensitive | |
Immiscible with water. | |
NC1=CC=C([S-])C=C1 | |
124.18 | |
125.19 | |
Crystalline |
Safety and Handling
GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302-H314
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, BASIC, ORGANIC, N.O.S.
missing translation for 'einecsNumber' : 214-763-4
missing translation for 'rtecsNumber' : DC0602500
missing translation for 'tsca' : No
missing translation for 'storageNote' : Air Sensitive
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only