missing translation for 'onlineSavingsMsg'
Learn More

4-Benzoyl-4'-bromobiphenyl, 99%, Thermo Scientific Chemicals

Product Code. 11448397
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63242-14-8
Molecular Formula C19H13BrO
Molecular Weight (g/mol) 337.216
MDL Number MFCD00209630
InChI Key ITMLYLPKSFZCJK-UHFFFAOYSA-N
Synonym 4-benzoyl-4'-bromobiphenyl, 4-4-bromophenyl benzophenone, 4'-bromobiphenyl-4-yl phenyl methanone, 4'-bromo-1,1'-biphenyl-4-yl phenyl methanone, 4-4-bromophenyl phenyl-phenyl-methanone, 4-4-bromophenyl phenyl phenyl ketone, 4-4-bromophenyl phenyl-phenylmethanone, 4-4-bromophenyl phenyl phenyl methanone, pubchem9089
PubChem CID 478901
IUPAC Name [4-(4-bromophenyl)phenyl]-phenylmethanone
SMILES C1=CC=C(C=C1)C(=O)C2=CC=C(C=C2)C3=CC=C(C=C3)Br

Specifications

Melting Point 155°C to 158°C
Quantity 500 g
Beilstein 2378429
Formula Weight 337.22
Percent Purity 99%
Chemical Name or Material 4-Benzoyl-4'-bromobiphenyl

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'