missing translation for 'onlineSavingsMsg'
Learn More

4-Benzyl-3-thiosemicarbazide, 98+%, Thermo Scientific Chemicals

Product Code. 11313076
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

4-Benzyl-3-thiosemicarbazide is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Benzyl-3-thiosemicarbazide is used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13431-41-9
Molecular Formula C8H11N3S
Molecular Weight (g/mol) 181.257
MDL Number MFCD00025145
InChI Key ZTRUHAVBRPABTK-UHFFFAOYSA-N
Synonym 4-benzyl-3-thiosemicarbazide, 4-benzylthiosemicarbazide, 3-amino-1-benzylthiourea, n-benzylhydrazinecarbothioamide, 4-benzylthiosemicarbazone, hydrazinecarbothioamide,n-phenylmethyl, hydrazinecarbothioamide, n-phenylmethyl, hydrazino benzylamino methane-1-thione, n-phenylmethyl hydrazinecarbothioamide, 1-amino-3-benzyl-thiourea
PubChem CID 737135
IUPAC Name 1-amino-3-benzylthiourea
SMILES C1=CC=C(C=C1)CNC(=S)NN

Specifications

Melting Point 128°C to 131°C
Quantity 1 g
UN Number UN2811
Beilstein 2096431
Solubility Information Insoluble in water.
Formula Weight 181.26
Percent Purity ≥98%
Chemical Name or Material 4-Benzyl-3-thiosemicarbazide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'