Learn More
4-Benzyloxy-2(1H)-pyridone, 98%, Thermo Scientific Chemicals
£41.13 - £157.00
Chemical Identifiers
CAS | 53937-02-3 |
---|---|
Molecular Formula | C12H11NO2 |
Molecular Weight (g/mol) | 201.23 |
MDL Number | MFCD04039771 |
InChI Key | DOVNUEPFPBWTSV-UHFFFAOYSA-N |
Synonym | 4-benzyloxy-2-1h-pyridone, 4-benzyloxy-2 1h-pyridone, 4-benzyloxy pyridin-2 1h-one, 4-benzyloxypyridin-2-one, 4-benzyloxy pyridin-2-ol, 4-benzyloxy-2-hydroxypyridine, 4-benzyloxy-1h-pyridin-2-one, 2 1h-pyridinone, 4-phenylmethoxy, 4-benzyloxy-1,2-dihydropyridin-2-one |
PubChem CID | 3607724 |
SMILES | O=C1NC=CC(OCC2=CC=CC=C2)=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10587790
|
Thermo Scientific Acros
438090010 |
1 g | |||||||
10587980
|
Thermo Scientific Acros
438090050 |
5 g |
£157.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
53937-02-3 | |
201.23 | |
DOVNUEPFPBWTSV-UHFFFAOYSA-N | |
3607724 |
C12H11NO2 | |
MFCD04039771 | |
4-benzyloxy-2-1h-pyridone, 4-benzyloxy-2 1h-pyridone, 4-benzyloxy pyridin-2 1h-one, 4-benzyloxypyridin-2-one, 4-benzyloxy pyridin-2-ol, 4-benzyloxy-2-hydroxypyridine, 4-benzyloxy-1h-pyridin-2-one, 2 1h-pyridinone, 4-phenylmethoxy, 4-benzyloxy-1,2-dihydropyridin-2-one | |
O=C1NC=CC(OCC2=CC=CC=C2)=C1 |
Specifications
53937-02-3 | |
100.0 | |
Brown to Yellow | |
97.5% min. (HPLC) | |
C12H11NO2 | |
4-benzyloxy-2-1h-pyridone, 4-benzyloxy-2 1h-pyridone, 4-benzyloxy pyridin-2 1h-one, 4-benzyloxypyridin-2-one, 4-benzyloxy pyridin-2-ol, 4-benzyloxy-2-hydroxypyridine, 4-benzyloxy-1h-pyridin-2-one, 2 1h-pyridinone, 4-phenylmethoxy, 4-benzyloxy-1,2-dihydropyridin-2-one | |
O=C1NC=CC(OCC2=CC=CC=C2)=C1 | |
201.23 | |
201.23 | |
Powder |
97.5 | |
199.7°C to 201.7°C | |
Authentic | |
Glass Bottle | |
MFCD04039771 | |
DOVNUEPFPBWTSV-UHFFFAOYSA-N | |
4-phenylmethoxy-1H-pyridin-2-one | |
3607724 | |
98% | |
4-Benzyloxy-2(1H)-pyridone |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: If breathing is difficult,remove to fresh air and keep at rest in a position comfortable for breathing.
GHS Signal Word: Warning
- 4-Benzyloxy-2(1H)-pyridone
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only