missing translation for 'onlineSavingsMsg'
Learn More

4-(Benzyloxy)-2-fluorophenylboronic acid, 97%, anhydride present, Thermo Scientific™

Product Code. 10782683
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 166744-78-1
Molecular Formula C13H12BFO3
Molecular Weight (g/mol) 246.044
MDL Number MFCD02093064
InChI Key PQWKGFALWCOVTC-UHFFFAOYSA-N
Synonym 4-benzyloxy-2-fluorophenylboronic acid, 4-benzyloxy-2-fluorobenzeneboronic acid, 4-benzyloxy-2-fluorophenyl boronic acid, boronic acid, 2-fluoro-4-phenylmethoxy phenyl, 2-fluoro-4-phenylmethoxyphenyl boronic acid, 4-benzyloxy-2-fluorophenylboronic acid, may contain varying amounts of anhydride, pubchem7760, akos brn-0077, acmc-209dv0
PubChem CID 2737786
IUPAC Name (2-fluoro-4-phenylmethoxyphenyl)boronic acid
SMILES B(C1=C(C=C(C=C1)OCC2=CC=CC=C2)F)(O)O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 246.04
Percent Purity 97%
Chemical Name or Material 4-(Benzyloxy)-2-fluorophenylboronic acid, May contain varying amounts of anhydride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'