missing translation for 'onlineSavingsMsg'
Learn More

4-Benzyloxy-3-methoxybenzeneboronic acid, 98%, Thermo Scientific™

Product Code. 15409207
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 243990-53-6
Molecular Formula C14H15BO4
Molecular Weight (g/mol) 258.08
MDL Number MFCD09027252
InChI Key PBVVDNNISDIUSU-UHFFFAOYSA-N
Synonym 4-benzyloxy-3-methoxyphenylboronic acid, 4-benzyloxy-3-methoxybenzeneboronic acid, 4-benzyloxy-3-methoxyphenyl boronic acid, 4-benzyloxoy-3-methoxyphenylboronic acid, acmc-209gbw, 4-benzyloxy-3-methoxyphenyl boronicacid, 4-benzyloxy-3-methoxybenzene boronic acid
PubChem CID 10682603
IUPAC Name (3-methoxy-4-phenylmethoxyphenyl)boronic acid
SMILES COC1=C(OCC2=CC=CC=C2)C=CC(=C1)B(O)O

Specifications

Melting Point 200°C to 203°C
Quantity 1 g
Formula Weight 258.08
Percent Purity 98%
Chemical Name or Material 4-Benzyloxy-3-methoxybenzeneboronic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'