Learn More
4-(Benzyloxy)phenol, 98%, Thermo Scientific Chemicals
£39.20 - £382.00
Chemical Identifiers
CAS | 103-16-2 |
---|---|
Molecular Formula | C13H12O2 |
Molecular Weight (g/mol) | 200.24 |
MDL Number | MFCD00002333 |
InChI Key | VYQNWZOUAUKGHI-UHFFFAOYSA-N |
Synonym | 4-benzyloxyphenol, monobenzone, 4-benzyloxy phenol, hydroquinone monobenzyl ether, benoquin, benzoquin, superlite, monobenzyl hydroquinone, leucodinine, monobenzon |
PubChem CID | 7638 |
ChEBI | CHEBI:34380 |
IUPAC Name | 4-phenylmethoxyphenol |
SMILES | C1=CC=C(C=C1)COC2=CC=C(C=C2)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10791631
|
Thermo Scientific Acros
169780500 |
50 g | Glass bottle |
£39.20
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10457010
|
Thermo Scientific Acros
169782500 |
250 g | Glass bottle |
£139.00
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
13498408
|
Thermo Scientific Acros
169780010 |
1 kg | Glass bottle |
£382.00
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
103-16-2 | |
200.24 | |
VYQNWZOUAUKGHI-UHFFFAOYSA-N | |
7638 | |
4-phenylmethoxyphenol |
C13H12O2 | |
MFCD00002333 | |
4-benzyloxyphenol, monobenzone, 4-benzyloxy phenol, hydroquinone monobenzyl ether, benoquin, benzoquin, superlite, monobenzyl hydroquinone, leucodinine, monobenzon | |
CHEBI:34380 | |
C1=CC=C(C=C1)COC2=CC=C(C=C2)O |
Specifications
103-16-2 | |
100.0 | |
Brown to White | |
97.5% min. (GC) | |
C6H5CH2OC6H4OH | |
06,845 | |
4-benzyloxyphenol, monobenzone, 4-benzyloxy phenol, hydroquinone monobenzyl ether, benoquin, benzoquin, superlite, monobenzyl hydroquinone, leucodinine, monobenzon | |
VYQNWZOUAUKGHI-UHFFFAOYSA-N | |
4-phenylmethoxyphenol | |
7638 | |
200.24 | |
Powder, Crystals or Chunks |
97.5 | |
121.0°C to 122.0°C | |
Authentic | |
C13H12O2 | |
MFCD00002333 | |
15,6334 | |
Solubility in water: slightly soluble. Other solubilities: very soluble in ethanol,benzene,diethyl ether,,soluble in acetone and alkalies,,practically insoluble in petroleum hydrocarbons | |
C1=CC=C(C=C1)COC2=CC=C(C=C2)O | |
200.24 | |
CHEBI:34380 | |
98% | |
4-(Benzyloxy)phenol, 98% |
Safety and Handling
GHS H Statement:
May cause an allergic skin reaction.
Causes serious eye irritation.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 203-083-3
missing translation for 'rtecsNumber' : SJ7700000
missing translation for 'tsca' : TSCA
- 4-Benzyloxyphenol
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Respiratory sensitiser Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
- H319-Causes serious eye irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only