missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Benzyloxyphenol, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
50 g
100 g
250 g
1000 g
Description
4-Benzyloxyphenol is used in the synthesis of bis(4-benzyloxyphenoxy)phenyl phosphine oxide. It plays an essential role in the preparation of hetaryl-azophenol dyes via heterocyclic amines in nitrosyl sulphuric acid. It is also used for polyester fiber dyeing and in rubber industry. It acts as a depigmenting agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Benzyloxyphenol is used in the synthesis of bis(4-benzyloxyphenoxy)phenyl phosphine oxide. It plays an essential role in the preparation of hetaryl-azophenol dyes via heterocyclic amines in nitrosyl sulphuric acid. It is also used for polyester fiber dyeing and in rubber industry. It acts as a depigmenting agent.
Solubility
Slightly soluble in water. Soluble in alcohol, acetone, benzene, and diethyl ether.
Notes
Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents and strong bases.
4-Benzyloxyphenol is used in the synthesis of bis(4-benzyloxyphenoxy)phenyl phosphine oxide. It plays an essential role in the preparation of hetaryl-azophenol dyes via heterocyclic amines in nitrosyl sulphuric acid. It is also used for polyester fiber dyeing and in rubber industry. It acts as a depigmenting agent.
Solubility
Slightly soluble in water. Soluble in alcohol, acetone, benzene, and diethyl ether.
Notes
Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents and strong bases.
Spécification
Spécification
Melting Point | 118°C to 122°C |
Density | 1.26 |
Quantity | 250 g |
Beilstein | 1958305 |
Merck Index | 14,6248 |
Solubility Information | Slightly soluble aq. soln.. Soluble in alcohol,acetone,benzene,and diethyl ether. |
SMILES | OC1=CC=C(OCC2=CC=CC=C2)C=C1 |
Molecular Weight (g/mol) | 200.24 |
Formula Weight | 200.24 |
Percent Purity | ≥98% |
Afficher plus |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'