missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Benzylphenol, Thermo Scientific™
Click to view available options
Quantity:
1 g
Specifications
Alpha Vector | BENZYLPHENOL |
Melting Point | 80°C to 85°C |
Color | White to beige to brown |
Infrared Spectrum | Conforms |
Packaging | Glass Bottle |
Linear Formula | C6H5CH2C6H4OH |
Quantity | 1 g |
Beilstein | 06-675 |
Merck Index | 15-1145 |
Physical Form | Crystalline Powder or Crystals |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'