missing translation for 'onlineSavingsMsg'
Learn More
4-Benzylpiperazine-1-carboximidamide hydroiodide, 95%, Thermo Scientific™ Product Code.: 11316895

4-Benzylpiperazine-1-carboximidamide hydroiodide, 95%, Thermo Scientific™

Product Code. 11316895
10 g, Amber Glass Bottle
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy

Product Code. 11316895

Brand: Thermo Scientific Maybridge MO07679EA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 852228-15-0
Molecular Formula C12H19IN4
Molecular Weight (g/mol) 346.22
MDL Number MFCD09025921
InChI Key CXQITJKGJPFOIV-UHFFFAOYSA-N
Synonym 4-benzylpiperazine-1-carboximidamide hydroiodide, 4-benzylpiperazine-1-carboximidamide-hydrogen iodide 1/1
PubChem CID 18525929
IUPAC Name 4-benzylpiperazine-1-carboximidamide;hydroiodide
SMILES I.NC(=N)N1CCN(CC2=CC=CC=C2)CC1

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 346.21
Percent Purity 95%
Chemical Name or Material 4-Benzylpiperazine-1-carboximidamide hydroiodide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Benzylpiperazine-1-carboximidamide hydroiodide, 95%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.