Learn More
4-Biphenyl isocyanate, 97%, Thermo Scientific Chemicals
£58.60 - £214.00
Chemical Identifiers
CAS | 92-95-5 |
---|---|
Molecular Formula | C13H9NO |
Molecular Weight (g/mol) | 195.22 |
MDL Number | MFCD00037089 |
InChI Key | WIRPZDICFIIBRF-UHFFFAOYSA-N |
Synonym | 4-biphenylyl isocyanate, p-xenylcarbimide, 4-biphenyl isocyanate, 4-isocyanatobiphenyl, 4-phenylphenyl isocyanate, unii-b0zsp4upq4, 4-biphenylylisocyanate, b0zsp4upq4, p-diphenyl isocyanate, p-xenylcarbimide mi |
PubChem CID | 3612319 |
IUPAC Name | 1-isocyanato-4-phenylbenzene |
SMILES | O=C=NC1=CC=C(C=C1)C1=CC=CC=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11612699
|
Thermo Scientific Acros
438500010 |
1 g |
£58.60
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11955691
|
Thermo Scientific Acros
438500050 |
5 g |
£214.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
92-95-5 | |
195.22 | |
WIRPZDICFIIBRF-UHFFFAOYSA-N | |
3612319 | |
O=C=NC1=CC=C(C=C1)C1=CC=CC=C1 |
C13H9NO | |
MFCD00037089 | |
4-biphenylyl isocyanate, p-xenylcarbimide, 4-biphenyl isocyanate, 4-isocyanatobiphenyl, 4-phenylphenyl isocyanate, unii-b0zsp4upq4, 4-biphenylylisocyanate, b0zsp4upq4, p-diphenyl isocyanate, p-xenylcarbimide mi | |
1-isocyanato-4-phenylbenzene |
Specifications
92-95-5 | |
100.0 | |
Brown to Orange | |
Authentic | |
Glass bottle | |
MFCD00037089 | |
4-biphenylyl isocyanate, p-xenylcarbimide, 4-biphenyl isocyanate, 4-isocyanatobiphenyl, 4-phenylphenyl isocyanate, unii-b0zsp4upq4, 4-biphenylylisocyanate, b0zsp4upq4, p-diphenyl isocyanate, p-xenylcarbimide mi | |
O=C=NC1=CC=C(C=C1)C1=CC=CC=C1 | |
195.22 | |
195.22 | |
Solution |
96.0 | |
60.0°C to 61.0°C | |
110.0°C to 112.0°C (1.0 mmHg) | |
96% min. (GC) | |
C13H9NO | |
15,10271 | |
WIRPZDICFIIBRF-UHFFFAOYSA-N | |
1-isocyanato-4-phenylbenzene | |
3612319 | |
97% | |
4-Biphenyl isocyanate |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye irritation.
Harmful if inhaled.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plen
GHS Signal Word: Danger
- 4-Biphenyl isocyanate, 97%
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
- Specific target organ toxicity Category 3
Hazard Statement
- H317-May cause an allergic skin reaction.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
RUO – Research Use Only