Learn More
4-Boc-1-(6-bromo-2-pyridyl)piperazine, 97%, Thermo Scientific™
£24.10 - £338.00
Chemical Identifiers
CAS | 331767-56-7 |
---|---|
Molecular Formula | C14H20BrN3O2 |
Molecular Weight (g/mol) | 342.237 |
MDL Number | MFCD07369773 |
InChI Key | CQSAPHUUCUUHNS-UHFFFAOYSA-N |
Synonym | 4-boc-1-6-bromo-2-pyridyl piperazine, tert-butyl 4-6-bromopyridin-2-yl piperazine-1-carboxylate, t-butyl 4-6-bromopyridin-2-yl piperazine-1-carboxylate, 2-n-boc-piperazin-1-yl-6-bromopyridine, 1-piperazinecarboxylic acid, 4-6-bromo-2-pyridinyl-, 1,1-dimethylethyl ester, acmc-1ctvy, 4-boc-1-6-bromo-pyridin-2-yl piperazine, tert-butyl 4-6'-bromopyrid-2'-yl piperazine-1-carboxylate, tert.butyl 4-6-bromo-pyridin-2-yl-piperazine-1-carboxylate |
PubChem CID | 16059046 |
IUPAC Name | tert-butyl 4-(6-bromopyridin-2-yl)piperazine-1-carboxylate |
SMILES | CC(C)(C)OC(=O)N1CCN(CC1)C2=NC(=CC=C2)Br |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
331767-56-7 | |
342.237 | |
CQSAPHUUCUUHNS-UHFFFAOYSA-N | |
16059046 | |
CC(C)(C)OC(=O)N1CCN(CC1)C2=NC(=CC=C2)Br |
C14H20BrN3O2 | |
MFCD07369773 | |
4-boc-1-6-bromo-2-pyridyl piperazine, tert-butyl 4-6-bromopyridin-2-yl piperazine-1-carboxylate, t-butyl 4-6-bromopyridin-2-yl piperazine-1-carboxylate, 2-n-boc-piperazin-1-yl-6-bromopyridine, 1-piperazinecarboxylic acid, 4-6-bromo-2-pyridinyl-, 1,1-dimethylethyl ester, acmc-1ctvy, 4-boc-1-6-bromo-pyridin-2-yl piperazine, tert-butyl 4-6'-bromopyrid-2'-yl piperazine-1-carboxylate, tert.butyl 4-6-bromo-pyridin-2-yl-piperazine-1-carboxylate | |
tert-butyl 4-(6-bromopyridin-2-yl)piperazine-1-carboxylate |
Specifications
331767-56-7 | |
White to Yellow | |
MFCD07369773 | |
4-boc-1-6-bromo-2-pyridyl piperazine, tert-butyl 4-6-bromopyridin-2-yl piperazine-1-carboxylate, t-butyl 4-6-bromopyridin-2-yl piperazine-1-carboxylate, 2-n-boc-piperazin-1-yl-6-bromopyridine, 1-piperazinecarboxylic acid, 4-6-bromo-2-pyridinyl-, 1,1-dimethylethyl ester, acmc-1ctvy, 4-boc-1-6-bromo-pyridin-2-yl piperazine, tert-butyl 4-6'-bromopyrid-2'-yl piperazine-1-carboxylate, tert.butyl 4-6-bromo-pyridin-2-yl-piperazine-1-carboxylate | |
CC(C)(C)OC(=O)N1CCN(CC1)C2=NC(=CC=C2)Br | |
342.237 | |
342.23 | |
Powder |
94°C to 98°C | |
C14H20BrN3O2 | |
UN2811 | |
CQSAPHUUCUUHNS-UHFFFAOYSA-N | |
tert-butyl 4-(6-bromopyridin-2-yl)piperazine-1-carboxylate | |
16059046 | |
97% | |
4-Boc-1-(6-bromo-2-pyridyl)piperazine |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H301-H315-H319-H335
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures