missing translation for 'onlineSavingsMsg'
Learn More

4-Boc-1-(6-bromo-2-pyridyl)piperazine, 97%, Thermo Scientific™

Product Code. 15441378
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 331767-56-7
Molecular Formula C14H20BrN3O2
Molecular Weight (g/mol) 342.237
MDL Number MFCD07369773
InChI Key CQSAPHUUCUUHNS-UHFFFAOYSA-N
Synonym 4-boc-1-6-bromo-2-pyridyl piperazine, tert-butyl 4-6-bromopyridin-2-yl piperazine-1-carboxylate, t-butyl 4-6-bromopyridin-2-yl piperazine-1-carboxylate, 2-n-boc-piperazin-1-yl-6-bromopyridine, 1-piperazinecarboxylic acid, 4-6-bromo-2-pyridinyl-, 1,1-dimethylethyl ester, acmc-1ctvy, 4-boc-1-6-bromo-pyridin-2-yl piperazine, tert-butyl 4-6'-bromopyrid-2'-yl piperazine-1-carboxylate, tert.butyl 4-6-bromo-pyridin-2-yl-piperazine-1-carboxylate
PubChem CID 16059046
IUPAC Name tert-butyl 4-(6-bromopyridin-2-yl)piperazine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCN(CC1)C2=NC(=CC=C2)Br

Specifications

Melting Point 94°C to 98°C
Color White to Yellow
Quantity 1 g
UN Number UN2811
Formula Weight 342.23
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 4-Boc-1-(6-bromo-2-pyridyl)piperazine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'