missing translation for 'onlineSavingsMsg'
Learn More

4-(Boc-amino)-2-chlorobenzoic acid, 97%, Thermo Scientific Chemicals

Product Code. 15489527
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 232275-73-9
Molecular Formula C12H14ClNO4
Molecular Weight (g/mol) 271.697
MDL Number MFCD02682164
InChI Key HDYBNYDKCZJPPX-UHFFFAOYSA-N
Synonym n-boc-4-amino-2-chlorobenzoic acid, 4-tert-butoxycarbonyl amino-2-chlorobenzoic acid, 2-chloro-4-boc-amino-benzoic acid, 4-tert-butoxy carbonyl amino-2-chlorobenzoic acid, 2-chloro-4-boc-amino benzoic acid, 4-tert-butoxycarbonylamino-2-chloro-benzoic acid, 4-tert-butoxycarbonylamino-2-chlorobenzoic acid, benzoic acid,2-chloro-4-1,1-dimethylethoxy carbonyl amino
PubChem CID 17750449
IUPAC Name 2-chloro-4-[(2-methylpropan-2-yl)oxycarbonylamino]benzoic acid
SMILES CC(C)(C)OC(=O)NC1=CC(=C(C=C1)C(=O)O)Cl

Specifications

Quantity 250 mg
Formula Weight 271.7
Percent Purity 97%
Chemical Name or Material 4-(Boc-amino)-2-chlorobenzoic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'