missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(Boc-amino)cyclohexanecarboxylic acid, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
4-(Boc-amino)cyclohexanecarboxylic acid is used as pharmaceutical intermediate.
Solubility
Soluble in methanol and tetrahydrofuran(THF).
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
4-(Boc-amino)cyclohexanecarboxylic acid is used as pharmaceutical intermediate.
Solubility
Soluble in methanol and tetrahydrofuran(THF).
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Solubility Information | Soluble in methanol and tetrahydrofuran(THF). |
Formula Weight | 243.3 |
Percent Purity | 97% |
Quantity | 1 g |
Chemical Name or Material | 4-(Boc-amino)cyclohexanecarboxylic acid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'