missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-1-butene, 98+%, Thermo Scientific Chemicals

Product Code. 10533351
Click to view available options
Quantity:
5 mL
25 mL
50 mL
Packaging:
Glass bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5162-44-7
Molecular Formula C4H7Br
Molecular Weight (g/mol) 135
MDL Number MFCD00000258
InChI Key DMAYBPBPEUFIHJ-UHFFFAOYSA-N
Synonym 4-bromo-1-butene, 3-butenyl bromide, 1-butene, 4-bromo, 4-bromo-1-buten, 4-bromo-but-1-ene, homoallyl bromide, 1-bromo-3-butene, 4-bromobutene, 4-bromo-1 butene, sjphabijuh@
PubChem CID 21241
IUPAC Name 4-bromobut-1-ene
SMILES C=CCCBr

Specifications

CAS Min % 98.0
CAS Max % 100.0
Color Brown to Colorless
Density 1.3300g/mL
Boiling Point 98.0°C to 100.0°C
Flash Point 1°C
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Linear Formula BrCH2CH2CH=CH2
Refractive Index 1.4595 to 1.4635
Quantity 25 mL
Beilstein 01, I, 84
Specific Gravity 1.33
Formula Weight 135
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material 4-Bromo-1-butene, 98+%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 4-Bromo-1-butene
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'