missing translation for 'onlineSavingsMsg'
Learn More

(4-Bromo-1-methyl-1H-pyrazol-3-yl)methanol, ≥97%, Thermo Scientific™

Product Code. 10265884
missing translation for 'orderingAttributeHoverText'
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 915707-65-2
Molecular Formula C5H7BrN2O
Molecular Weight (g/mol) 191.028
MDL Number MFCD06202888
InChI Key JFGLJTFTVBZOCB-UHFFFAOYSA-N
Synonym 4-bromo-1-methyl-1h-pyrazol-3-yl methanol, 4-bromo-1-methylpyrazol-3-yl methanol, 4-bromo-3-hydroxymethyl-1-methyl-1h-pyrazole, 4-bromo-1-methylpyrazol-3-yl methan-1-ol, pubchem23717
PubChem CID 24229713
IUPAC Name (4-bromo-1-methylpyrazol-3-yl)methanol
SMILES CN1C=C(C(=N1)CO)Br

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97+%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 191.03
Percent Purity 97%
Chemical Name or Material (4-Bromo-1-methyl-1H-pyrazol-3-yl)methanol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'