missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-1-methyl-1H-pyrazole-3-carbonitrile, 97%, Thermo Scientific™

Product Code. 10097879
Click to view available options
Quantity:
1 g
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 10097879

Brand: Thermo Scientific Maybridge MAY00324DA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 287922-71-8
Assay Percent Range 97%
Molecular Formula C5H4BrN3
MDL Number MFCD00103545
Quantity 1 g
Synonym 4-bromo-1-methyl-1h-pyrazole-3-carbonitrile, 1h-pyrazole-3-carbonitrile, 4-bromo-1-methyl, 4-bromo-1-methyl-pyrazole-3-carbonitrile
InChI Key NTTLCOOFUGHMJS-UHFFFAOYSA-N
SMILES CN1C=C(Br)C(=N1)C#N
IUPAC Name 4-bromo-1-methylpyrazole-3-carbonitrile
Molecular Weight (g/mol) 186.01
PubChem CID 45594322
Formula Weight 186.01
Chemical Name or Material 4-Bromo-1-methyl-1H-pyrazole-3-carbonitrile
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.