missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-1-methyl-1H-pyrazole-3-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10327364
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 84547-86-4
Molecular Formula C5H5BrN2O2
Molecular Weight (g/mol) 205.011
InChI Key LEEPGDCCHVRYHK-UHFFFAOYSA-N
Synonym 4-bromo-1-methyl-1h-pyrazole-3-carboxylic acid, 1h-pyrazole-3-carboxylic acid, 4-bromo-1-methyl, 4-bromo-1-methyl-1h-pyrazole-3-carboxylicacid, pubchem16814, 4-bromo-1-methyl-pyrazole-3-carboxylic acid, 1h-pyrazole-3-carboxylicacid, 4-bromo-1-methyl, 4-bromo-1-methyl-1h-pyrazole-3-carboxylic acid #
PubChem CID 536013
IUPAC Name 4-bromo-1-methylpyrazole-3-carboxylic acid
SMILES CN1C=C(C(=N1)C(=O)O)Br

Specifications

Assay Percent Range 97%
Quantity 250 mg
Formula Weight 205.01
Percent Purity 97%
Chemical Name or Material 4-Bromo-1-methyl-1H-pyrazole-3-carboxylic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.