missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Bromo-1H-pyrazole-5-carbaldehyde, 95+%, Thermo Scientific™
£54.10 - £266.00
Chemical Identifiers
CAS | 287917-97-9 |
---|---|
Molecular Formula | C4H3BrN2O |
Molecular Weight (g/mol) | 174.985 |
InChI Key | UWGFONONBAIDAF-UHFFFAOYSA-N |
Synonym | 4-bromo-1h-pyrazole-3-carbaldehyde, 4-bromo-1h-pyrazole-5-carboxaldehyde, 4-bromo-2h-pyrazole-3-carbaldehyde, 4-bromo-1h-pyrazole-3-carboxaldehyde, 1h-pyrazole-3-carboxaldehyde,4-bromo, 1h-pyrazole-3-carboxaldehyde, 4-bromo, 4-bromopyrazole-5-carbaldehyde, 4-bromo-5-formyl-1h-pyrazole, 4-bromo-pyrazole-3-carboxaldehyde, 4-bromo-2h-pyrazole-3-carboxaldehyde |
PubChem CID | 2782154 |
IUPAC Name | 4-bromo-1H-pyrazole-5-carbaldehyde |
SMILES | C1=NNC(=C1Br)C=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10550353
|
Thermo Scientific Maybridge
RF03950CB |
250 mg |
£54.10
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10344202
|
Thermo Scientific Maybridge
RF03950DA |
1 g |
£137.00
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10442654
|
Thermo Scientific Maybridge
RF03950DE |
5 g |
£266.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
287917-97-9 | |
174.985 | |
4-bromo-1h-pyrazole-3-carbaldehyde, 4-bromo-1h-pyrazole-5-carboxaldehyde, 4-bromo-2h-pyrazole-3-carbaldehyde, 4-bromo-1h-pyrazole-3-carboxaldehyde, 1h-pyrazole-3-carboxaldehyde,4-bromo, 1h-pyrazole-3-carboxaldehyde, 4-bromo, 4-bromopyrazole-5-carbaldehyde, 4-bromo-5-formyl-1h-pyrazole, 4-bromo-pyrazole-3-carboxaldehyde, 4-bromo-2h-pyrazole-3-carboxaldehyde | |
4-bromo-1H-pyrazole-5-carbaldehyde |
C4H3BrN2O | |
UWGFONONBAIDAF-UHFFFAOYSA-N | |
2782154 | |
C1=NNC(=C1Br)C=O |
Specifications
287917-97-9 | |
C4H3BrN2O | |
UWGFONONBAIDAF-UHFFFAOYSA-N | |
4-bromo-1H-pyrazole-5-carbaldehyde | |
2782154 | |
95% |
95% | |
4-bromo-1h-pyrazole-3-carbaldehyde, 4-bromo-1h-pyrazole-5-carboxaldehyde, 4-bromo-2h-pyrazole-3-carbaldehyde, 4-bromo-1h-pyrazole-3-carboxaldehyde, 1h-pyrazole-3-carboxaldehyde,4-bromo, 1h-pyrazole-3-carboxaldehyde, 4-bromo, 4-bromopyrazole-5-carbaldehyde, 4-bromo-5-formyl-1h-pyrazole, 4-bromo-pyrazole-3-carboxaldehyde, 4-bromo-2h-pyrazole-3-carboxaldehyde | |
C1=NNC(=C1Br)C=O | |
174.985 | |
174.98 | |
4-Bromo-1H-pyrazole-5-carbaldehyde |