missing translation for 'onlineSavingsMsg'
Learn More
4-Bromo-1H-pyrazole, ≥98%, Thermo Scientific Chemicals Product Code.: 15421979

4-Bromo-1H-pyrazole, ≥98%, Thermo Scientific Chemicals

Product Code. 15421979
25 g
Click to view available options
Quantity:
25 g
500 g
1 kg
This item is not returnable. View return policy

Product Code. 15421979

Brand: Thermo Scientific Alfa Aesar H64673.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

4-Bromopyrazole may be used in the preparation of 4-bromo-1-(2-chloroethyl)-1H-pyrazole. It may be used as starting material in the synthesis of 1,4?-bipyrazoles. Also used in the synthesis of various pharmaceutical and biologically active compounds, including inhibitors. It is also shown to be one of the 4-substituted pyrazoles, being able to act as inhibitor of liver alcohol dehydrogenase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromopyrazole may be used in the preparation of 4-bromo-1-(2-chloroethyl)-1H-pyrazole. It may be used as starting material in the synthesis of 1,4′-bipyrazoles. Also used in the synthesis of various pharmaceutical and biologically active compounds, including inhibitors. It is also shown to be one of the 4-substituted pyrazoles, being able to act as inhibitor of liver alcohol dehydrogenase.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2075-45-8
Molecular Formula C3H3BrN2
Molecular Weight (g/mol) 146.98
MDL Number MFCD00075602
InChI Key WVGCPEDBFHEHEZ-UHFFFAOYSA-N
Synonym 4-bromopyrazole, 1h-pyrazole, 4-bromo, 4-brompyrazole, 4-bromo-pyrazole, pyrazole, 4-bromo, 4-bromo pyrazole, byz, zlchem 126, 1h-pyrazole,4-bromo, pubchem11041
PubChem CID 16375
IUPAC Name 4-bromo-1H-pyrazole
SMILES BrC1=CNN=C1

Specifications

Melting Point 93°C to 96°C (literature)
Boiling Point 250°C to 260°C (literature)
Quantity 25 g
Beilstein 106510
Solubility Information Slightly soluble in water.
Formula Weight 146.97
Percent Purity ≥98%
Physical Form Solid
Chemical Name or Material 4-Bromo-1H-pyrazole

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberN06J054 Certificate TypeCertificate of Analysis Date20/02/2024 Catalog NumberH64673.14,H64673.36,H64673.A1
Lot NumberN16J015 Certificate TypeCertificate of Analysis Date09/05/2023 Catalog NumberH64673.14,H64673.36
Lot NumberT06I051 Certificate TypeCertificate of Analysis Date13/07/2022 Catalog NumberH64673.14,H64673.36
Lot NumberY02G007 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberH64673.14,H64673.36
Lot Number81302128 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH64673.14,H64673.36

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Bromo-1H-pyrazole, ≥98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.