missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4'-Bromo-2,2,2-trifluoroacetophenone, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
4?-Bromo-2,2,2-trifluoroacetophenone may be used in the preparation of carbonyl-bridged bithiazole derivatives. Also used as a reagent to synthesize MK-5046, a selective Bombesin receptor subtype-3 agonist used to treat obesity.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4′-Bromo-2,2,2-trifluoroacetophenone may be used in the preparation of carbonyl-bridged bithiazole derivatives. Also used as a reagent to synthesize MK-5046, a selective Bombesin receptor subtype-3 agonist used to treat obesity.
Solubility
Slightly soluble in water.
Notes
Moisture Sensitive. Store away from water/moisture. Store away from oxidizing agent.
4′-Bromo-2,2,2-trifluoroacetophenone may be used in the preparation of carbonyl-bridged bithiazole derivatives. Also used as a reagent to synthesize MK-5046, a selective Bombesin receptor subtype-3 agonist used to treat obesity.
Solubility
Slightly soluble in water.
Notes
Moisture Sensitive. Store away from water/moisture. Store away from oxidizing agent.
Specifications
Specifications
Melting Point | 26°C to 30°C |
Boiling Point | 95°C (4 mmHg) |
Quantity | 1 g |
Sensitivity | Moisture sensitive |
Solubility Information | Slightly soluble in water. |
Formula Weight | 253.02 |
Percent Purity | 97% |
Chemical Name or Material | 4'-Bromo-2,2,2-trifluoroacetophenone |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'