missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Bromo-2,6-dimethylphenol, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
500 g
Description
It is used as a preservatives for coatings, slurries and to control microbial fouling in paper mills , oil field and leather process and water treatment process against microbial attack.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used as a preservatives for coatings, slurries and to control microbial fouling in paper mills , oil field and leather process and water treatment process against microbial attack.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
It is used as a preservatives for coatings, slurries and to control microbial fouling in paper mills , oil field and leather process and water treatment process against microbial attack.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications
Specifications
Melting Point | 74°C to 78°C |
Odor | Phenol-like |
Quantity | 100 g |
Beilstein | 1862399 |
Solubility Information | Slightly soluble in water. |
Formula Weight | 201.07 |
Percent Purity | 99% |
Chemical Name or Material | 4-Bromo-2,6-dimethylphenol |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'