missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-chloro-6-methylphenol, 97%, Thermo Scientific™

£8.10 - £117.00

Chemical Identifiers

CAS 7530-27-0
Molecular Formula C7H6BrClO
Molecular Weight (g/mol) 221.48
MDL Number MFCD00002167
InChI Key IDDUDPYBPXKGCP-UHFFFAOYSA-N
PubChem CID 82031
IUPAC Name 4-bromo-2-chloro-6-methylphenol
SMILES CC1=CC(Br)=CC(Cl)=C1O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10039561
View Documents
Thermo Scientific Maybridge
SB01216DA
1 g
£8.10
1g
Please to purchase this item. Need a web account? Register with us today!
10223662
View Documents
Thermo Scientific Maybridge
SB01216EA
10 g
£40.30
10g
Please to purchase this item. Need a web account? Register with us today!
10435712
View Documents
Thermo Scientific Maybridge
SB01216EE
50 g
£117.00
50g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

7530-27-0
221.48
IDDUDPYBPXKGCP-UHFFFAOYSA-N
4-bromo-2-chloro-6-methylphenol
C7H6BrClO
MFCD00002167
82031
CC1=CC(Br)=CC(Cl)=C1O

Specifications

7530-27-0
97%
C7H6BrClO
MFCD00002167
IDDUDPYBPXKGCP-UHFFFAOYSA-N
4-bromo-2-chloro-6-methylphenol
82031
97%
97.0
Amber Glass Bottle
CH3C6H2(OH)(Cl)Br
4-bromo-6-chloro-o-cresol, o-cresol, 4-bromo-6-chloro, 4-bromo-6-chloro-2-methylphenol, phenol, 4-bromo-2-chloro-6-methyl, 2-chloro-4-bromo-6-methylphenol, 4-bromo-2-chloro-6-methyl-phenol, 5-bromo-3-chloro-2-hydroxytoluene, #, phenol,4-bromo-2-chloro-6-methyl
CC1=CC(Br)=CC(Cl)=C1O
221.48
221.48
4-Bromo-2-chloro-6-methylphenol
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents

Product Certifications