missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-chlorobenzoic acid, 98+%, Thermo Scientific Chemicals

Product Code. 11404533
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59748-90-2
Molecular Formula C7H4BrClO2
Molecular Weight (g/mol) 235.461
MDL Number MFCD00040903
InChI Key JAVZWSOFJKYSDY-UHFFFAOYSA-N
Synonym benzoic acid, 4-bromo-2-chloro, 4-bromo-2-chlorobenzoicacid, 2-chloro-4-bromobenzoic acid, 4-bromo-2-chloro-benzoic acid, rarechem al bo 2371, attercop-chm at111793, zlchem 410, pubchem3586, acmc-1b03i, ksc274c2d
PubChem CID 108829
IUPAC Name 4-bromo-2-chlorobenzoic acid
SMILES C1=CC(=C(C=C1Br)Cl)C(=O)O

Specifications

Melting Point 172°C to 174°C
Quantity 5 g
Formula Weight 235.47
Percent Purity ≥98%
Chemical Name or Material 4-Bromo-2-chlorobenzoic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'