Learn More
4-Bromo-2-chlorobenzoic acid, 98+%, Thermo Scientific Chemicals
£13.40 - £116.00
Chemical Identifiers
CAS | 59748-90-2 |
---|---|
Molecular Formula | C7H4BrClO2 |
Molecular Weight (g/mol) | 235.461 |
MDL Number | MFCD00040903 |
InChI Key | JAVZWSOFJKYSDY-UHFFFAOYSA-N |
Synonym | benzoic acid, 4-bromo-2-chloro, 4-bromo-2-chlorobenzoicacid, 2-chloro-4-bromobenzoic acid, 4-bromo-2-chloro-benzoic acid, rarechem al bo 2371, attercop-chm at111793, zlchem 410, pubchem3586, acmc-1b03i, ksc274c2d |
PubChem CID | 108829 |
IUPAC Name | 4-bromo-2-chlorobenzoic acid |
SMILES | C1=CC(=C(C=C1Br)Cl)C(=O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11494523
|
Thermo Scientific Alfa Aesar
A11527.03 |
1 g | |||||||
11404533
|
Thermo Scientific Alfa Aesar
A11527.06 |
5 g |
£40.50
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11414533
|
Thermo Scientific Alfa Aesar
A11527.14 |
25 g |
£116.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
59748-90-2 | |
235.461 | |
JAVZWSOFJKYSDY-UHFFFAOYSA-N | |
108829 | |
C1=CC(=C(C=C1Br)Cl)C(=O)O |
C7H4BrClO2 | |
MFCD00040903 | |
benzoic acid, 4-bromo-2-chloro, 4-bromo-2-chlorobenzoicacid, 2-chloro-4-bromobenzoic acid, 4-bromo-2-chloro-benzoic acid, rarechem al bo 2371, attercop-chm at111793, zlchem 410, pubchem3586, acmc-1b03i, ksc274c2d | |
4-bromo-2-chlorobenzoic acid |
Specifications
59748-90-2 | |
C7H4BrClO2 | |
benzoic acid, 4-bromo-2-chloro, 4-bromo-2-chlorobenzoicacid, 2-chloro-4-bromobenzoic acid, 4-bromo-2-chloro-benzoic acid, rarechem al bo 2371, attercop-chm at111793, zlchem 410, pubchem3586, acmc-1b03i, ksc274c2d | |
C1=CC(=C(C=C1Br)Cl)C(=O)O | |
235.461 | |
235.47 | |
4-Bromo-2-chlorobenzoic acid |
172°C to 174°C | |
MFCD00040903 | |
JAVZWSOFJKYSDY-UHFFFAOYSA-N | |
4-bromo-2-chlorobenzoic acid | |
108829 | |
≥98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H301-H315-H319-H335
missing translation for 'dotInformation' : Hazard Class: 6.1; Packaging Group: III
missing translation for 'einecsNumber' : 261-911-9
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only