missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-chlorophenol, 99%, Thermo Scientific Chemicals

£19.40 - £212.00

Chemical Identifiers

CAS 3964-56-5
Molecular Formula C6H4BrClO
Molecular Weight (g/mol) 207.45
MDL Number MFCD00002166
InChI Key VIBJPUXLAKVICD-UHFFFAOYSA-N
Synonym 2-chloro-4-bromophenol, phenol, 4-bromo-2-chloro, phenol, 4-bromochloro, unii-994sj34d8p, 4-bromo-2-chloro-phenol, attercop-chm at106179, 4-bromo-2-chlorophenol 25gr, 4-bromo-chlorophenol, pubchem3596, acmc-209j6p
PubChem CID 19859
ChEBI CHEBI:38852
IUPAC Name 4-bromo-2-chlorophenol
SMILES OC1=CC=C(Br)C=C1Cl
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10433975
View Documents
Thermo Scientific Acros
173530050
5 g Glass bottle
£19.40
5g
Please to purchase this item. Need a web account? Register with us today!
10448320
View Documents
Thermo Scientific Acros
173530250
25 g Glass bottle
£65.20
25g
Please to purchase this item. Need a web account? Register with us today!
10523161
View Documents
Thermo Scientific Acros
173531000
100 g Glass bottle
£212.00
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

3964-56-5
207.45
VIBJPUXLAKVICD-UHFFFAOYSA-N
19859
4-bromo-2-chlorophenol
C6H4BrClO
MFCD00002166
2-chloro-4-bromophenol, phenol, 4-bromo-2-chloro, phenol, 4-bromochloro, unii-994sj34d8p, 4-bromo-2-chloro-phenol, attercop-chm at106179, 4-bromo-2-chlorophenol 25gr, 4-bromo-chlorophenol, pubchem3596, acmc-209j6p
CHEBI:38852
OC1=CC=C(Br)C=C1Cl

Specifications

3964-56-5
100.0
White
>112°C
98.5% min. (GC)
BrC6H3(Cl)OH
06, II, 187
VIBJPUXLAKVICD-UHFFFAOYSA-N
4-bromo-2-chlorophenol
19859
207.45
Low Melting Mass or Crystalline Needles, Powder and Chunks
98.5
47°C to 51°C
232°C to 235°C
Authentic
C6H4BrClO
MFCD00002166
2-chloro-4-bromophenol, phenol, 4-bromo-2-chloro, phenol, 4-bromochloro, unii-994sj34d8p, 4-bromo-2-chloro-phenol, attercop-chm at106179, 4-bromo-2-chlorophenol 25gr, 4-bromo-chlorophenol, pubchem3596, acmc-209j6p
OC1=CC=C(Br)C=C1Cl
207.45
CHEBI:38852
99%
4-Bromo-2-chlorophenol, 99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Toxic if swallowed.
Causes skin irritation.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 223-572-5

Product Identifier
  • 4-Bromo-2-chlorophenol

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only