Learn More
4-Bromo-2-ethylbenzene-1-sulfonyl chloride, ≥95%, Thermo Scientific™
£24.20 - £174.00
Chemical Identifiers
CAS | 175278-24-7 |
---|---|
Molecular Formula | C8H8BrClO2S |
Molecular Weight (g/mol) | 283.564 |
MDL Number | MFCD00204178 |
InChI Key | UOIVESXBURLTPX-UHFFFAOYSA-N |
Synonym | 4-bromo-2-ethylbenzene-1-sulfonyl chloride, 4-bromo-2-ethylbenzene-1-sulfonylchloride, 4-bromo-2-ethylbenzenesulphonyl chloride, benzenesulfonylchloride, 4-bromo-2-ethyl, pubchem5062, 4-bromo-2-ethyl-benzenesulfonyl chloride, 4-bromo-2-ethylphenylsulfonyl chloride, 4-bromanyl-2-ethyl-benzenesulfonyl chloride, 4-bromo-2-ethyl-benzene-1-sulfonyl chloride, 4-bromo-2-ethylbenzene-1-sulphonyl chloride |
PubChem CID | 2737536 |
IUPAC Name | 4-bromo-2-ethylbenzenesulfonyl chloride |
SMILES | CCC1=C(C=CC(=C1)Br)S(=O)(=O)Cl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10621893
|
Thermo Scientific Maybridge
TL00690DA |
1 g | Amber Glass Bottle |
£24.20
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10395152
|
Thermo Scientific Maybridge
TL00690EA |
10 g | Amber Glass Bottle |
£174.00
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
175278-24-7 | |
283.564 | |
UOIVESXBURLTPX-UHFFFAOYSA-N | |
2737536 | |
CCC1=C(C=CC(=C1)Br)S(=O)(=O)Cl |
C8H8BrClO2S | |
MFCD00204178 | |
4-bromo-2-ethylbenzene-1-sulfonyl chloride, 4-bromo-2-ethylbenzene-1-sulfonylchloride, 4-bromo-2-ethylbenzenesulphonyl chloride, benzenesulfonylchloride, 4-bromo-2-ethyl, pubchem5062, 4-bromo-2-ethyl-benzenesulfonyl chloride, 4-bromo-2-ethylphenylsulfonyl chloride, 4-bromanyl-2-ethyl-benzenesulfonyl chloride, 4-bromo-2-ethyl-benzene-1-sulfonyl chloride, 4-bromo-2-ethylbenzene-1-sulphonyl chloride | |
4-bromo-2-ethylbenzenesulfonyl chloride |
Specifications
175278-24-7 | |
95% | |
MFCD00204178 | |
UOIVESXBURLTPX-UHFFFAOYSA-N | |
4-bromo-2-ethylbenzenesulfonyl chloride | |
2737536 | |
95% |
95.0 | |
C8H8BrClO2S | |
4-bromo-2-ethylbenzene-1-sulfonyl chloride, 4-bromo-2-ethylbenzene-1-sulfonylchloride, 4-bromo-2-ethylbenzenesulphonyl chloride, benzenesulfonylchloride, 4-bromo-2-ethyl, pubchem5062, 4-bromo-2-ethyl-benzenesulfonyl chloride, 4-bromo-2-ethylphenylsulfonyl chloride, 4-bromanyl-2-ethyl-benzenesulfonyl chloride, 4-bromo-2-ethyl-benzene-1-sulfonyl chloride, 4-bromo-2-ethylbenzene-1-sulphonyl chloride | |
CCC1=C(C=CC(=C1)Br)S(=O)(=O)Cl | |
283.564 | |
283.57 | |
4-Bromo-2-ethylbenzene-1-sulfonyl chloride |
Safety and Handling
GHS H Statement:
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger