missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Bromo-2-fluorobenzonitrile, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
4-Bromo-2-fluorobenzonitrile is used as a OLED intermediate, Pharmaceutical, electronic and chemical intermediate. It is used in the synthesis of heterocycles and liquid crystals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Bromo-2-fluorobenzonitrile is used as a OLED intermediate, Pharmaceutical, electronic and chemical intermediate. It is used in the synthesis of heterocycles and liquid crystals.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
4-Bromo-2-fluorobenzonitrile is used as a OLED intermediate, Pharmaceutical, electronic and chemical intermediate. It is used in the synthesis of heterocycles and liquid crystals.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
Specifications
Melting Point | 69°C to 72°C |
Boiling Point | 238°C to 239°C |
Quantity | 25 g |
UN Number | UN3439 |
Beilstein | 4231143 |
Solubility Information | Insoluble in water. |
Formula Weight | 200.01 |
Percent Purity | 99% |
Chemical Name or Material | 4-Bromo-2-fluorobenzonitrile |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'