missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-methoxybenzyl alcohol, 97%, Thermo Scientific™

Product Code. 11332439
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is used in azaquinone-methide-mediated depolymerization of aromatic carbamate oligomers.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17102-63-5
Molecular Formula C8H9BrO2
Molecular Weight (g/mol) 217.06
MDL Number MFCD07369762
InChI Key HBEIHPSICGGZIF-UHFFFAOYSA-N
PubChem CID 15128243
IUPAC Name (4-bromo-2-methoxyphenyl)methanol
SMILES COC1=C(CO)C=CC(Br)=C1

Specifications

Melting Point 66°C to 70°C
Quantity 1 g
Synonym 4-bromo-2-methoxybenzyl alcohol, 4-bromo-2-methoxyphenyl methanol, 4-bromo-2-methoxy-phenyl-methanol, benzenemethanol, 4-bromo-2-methoxy, acmc-209e2a, 2-methoxy-4-bromobenzyl alcohol, benzenemethanol,4-bromo-2-methoxy
Solubility Information Insoluble in water.
Formula Weight 217.06
Percent Purity 97%
Chemical Name or Material 4-Bromo-2-methoxybenzyl alcohol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.