missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Bromo-2-methoxypyridine, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
4-Bromo-2-methoxypyridine used in the synhesis of Imidazo [1, 2-a] pyrimidines. Suzuki coupling between 4-methoxyphenylboronic acid and 4-bromo-2-methoxypyridine affords 1, 5-disubstituted pyridones. Starting material used in a synthesis of crown-ester-bipyridines and viologens via sodium or nickel reductive coupling, side chain oxidation and esterification.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Bromo-2-methoxypyridine used in the synhesis of Imidazo [1, 2-a] pyrimidines. Suzuki coupling between 4-methoxyphenylboronic acid and 4-bromo-2-methoxypyridine affords 1, 5-disubstituted pyridones. Starting material used in a synthesis of crown-ester-bipyridines and viologens via sodium or nickel reductive coupling, side chain oxidation and esterification.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, heat.
4-Bromo-2-methoxypyridine used in the synhesis of Imidazo [1, 2-a] pyrimidines. Suzuki coupling between 4-methoxyphenylboronic acid and 4-bromo-2-methoxypyridine affords 1, 5-disubstituted pyridones. Starting material used in a synthesis of crown-ester-bipyridines and viologens via sodium or nickel reductive coupling, side chain oxidation and esterification.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, heat.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 25°C to 26°C |
Color | White to Yellow |
Boiling Point | 87°C (15 mmHg) |
Quantity | 5 g |
Beilstein | 115199 |
Solubility Information | Slightly soluble in water. |
Formula Weight | 188.02 |
Percent Purity | 98% |
Physical Form | Low Melting Solid |
Chemical Name or Material | 4-Bromo-2-methoxypyridine |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'