Learn More
4-Bromo-2-methylbenzoic acid, 97%, Thermo Scientific Chemicals
£58.70 - £188.00
Chemical Identifiers
CAS | 68837-59-2 |
---|---|
Molecular Formula | C8H7BrO2 |
Molecular Weight (g/mol) | 215.05 |
InChI Key | RVCJOGNLYVNRDN-UHFFFAOYSA-N |
Synonym | 2-methyl-4-bromobenzoic acid, 4-bromo-o-toluic acid, 4-bromo-2-methyl-benzoic acid, benzoic acid, 4-bromo-2-methyl, synquest 2721-9-x6, 4-bromo-2-methylbenz, 4-bromo-2-methylbenzoicacid, pubchem4029, 5-bromo-2-carboxytoluene, acmc-1baz5 |
PubChem CID | 99570 |
IUPAC Name | 4-bromo-2-methylbenzoic acid |
SMILES | CC1=C(C=CC(=C1)Br)C(=O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15549343
|
Thermo Scientific Acros
443400050 |
5 g |
£58.70
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15559343
|
Thermo Scientific Acros
443400250 |
25 g |
£188.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
68837-59-2 | |
215.05 | |
2-methyl-4-bromobenzoic acid, 4-bromo-o-toluic acid, 4-bromo-2-methyl-benzoic acid, benzoic acid, 4-bromo-2-methyl, synquest 2721-9-x6, 4-bromo-2-methylbenz, 4-bromo-2-methylbenzoicacid, pubchem4029, 5-bromo-2-carboxytoluene, acmc-1baz5 | |
4-bromo-2-methylbenzoic acid |
C8H7BrO2 | |
RVCJOGNLYVNRDN-UHFFFAOYSA-N | |
99570 | |
CC1=C(C=CC(=C1)Br)C(=O)O |
Specifications
68837-59-2 | |
Authentic | |
C8H7BrO2 | |
RVCJOGNLYVNRDN-UHFFFAOYSA-N | |
4-bromo-2-methylbenzoic acid | |
99570 | |
97% |
182.0°C to 184.0°C | |
96% min. (HPLC) | |
2-methyl-4-bromobenzoic acid, 4-bromo-o-toluic acid, 4-bromo-2-methyl-benzoic acid, benzoic acid, 4-bromo-2-methyl, synquest 2721-9-x6, 4-bromo-2-methylbenz, 4-bromo-2-methylbenzoicacid, pubchem4029, 5-bromo-2-carboxytoluene, acmc-1baz5 | |
CC1=C(C=CC(=C1)Br)C(=O)O | |
215.05 | |
215.05 | |
4-Bromo-2-methylbenzoic acid |
Safety and Handling
GHS H Statement Toxic if swallowed. Causes serious eye irritation.
GHS P Statement Wear eye protection/face protection. Wash face, hands and any exposed skin thoroughly after handling. IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. If eye irritation persists: Get medical advice/attention.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 272-437-7
- 4-Bromo-2-methylbenzoic acid
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H319-Causes serious eye irritation.
- H301-Toxic if swallowed.
Precautionary Statement
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P337+P313-If eye irritation persists: Get medical advice/attention.
RUO – Research Use Only